Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Catalog number FSA396982500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS258865-48-4
IUPAC Name[7-(dihydroxyboranyl)-9,9-dioctyl-9H-fluoren-2-yl]boronic acid
Molecular FormulaC29H44B2O4
InChI KeyHURJMQMZDPOUOU-UHFFFAOYSA-N
SMILESCCCCCCCCC1(CCCCCCCC)C2=CC(=CC=C2C2=C1C=C(C=C2)B(O)O)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Color)White to off-white