Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Catalog number FSA396980025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Specifications
Chemical Name or MaterialIsopropanol
Melting Point-89.5°C
Absorbance0.01 max. (EurPh) at 310nm, 0.02 max. (EurPh) at 290nm, 0.03 max. (EurPh) at 270nm, 0.1 max. (EurPh) at 250nm, 0.3 max. (EurPh) at 230nm
CAS67-63-0
ColorBrown to Purple