Pyridine, for HPLC
Pyridine, for HPLC
Pyridine, for HPLC
Thermo Scientific Chemicals
Pyridine, for HPLC
CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3968000101 LGlass Bottle
Catalog number FSA396800010
Price (MYR)
1,481.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,481.00
Add to cart
Chemical Identifiers
CAS307-30-2
IUPAC Name2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-ol
Molecular FormulaC8H3F15O
InChI KeyPJDOLCGOTSNFJM-UHFFFAOYSA-N
SMILESOCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline mass
GC>=97.5 %
Appearance (Color)White
Melting point43°C to 47°C
For HPLC
RUO – Research Use Only