Potassium hydroxide, ca. 50% solution in water
Potassium hydroxide, ca. 50% solution in water
Potassium hydroxide, ca. 50% solution in water
Potassium hydroxide, ca. 50% solution in water
Thermo Scientific Chemicals
Potassium hydroxide, ca. 50% solution in water
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3960600252.5 kgPlastic bottle
Catalog number FSA396060025
Price (MYR)
514.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
514.00
Add to cart
Chemical Identifiers
CAS4394-85-8
IUPAC Namemorpholine-4-carbaldehyde
Molecular FormulaC5H9NO2
InChI KeyLCEDQNDDFOCWGG-UHFFFAOYSA-N
SMILESO=CN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4850 to 1.4880 (20°C, 589 nm)
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless to yellow