2-Chloronicotinoyl chloride, 97%, Thermo Scientific Chemicals
2-Chloronicotinoyl chloride, 97%, Thermo Scientific Chemicals
2-Chloronicotinoyl chloride, 97%, Thermo Scientific Chemicals
2-Chloronicotinoyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chloronicotinoyl chloride, 97%, Thermo Scientific Chemicals
CAS: 49609-84-9 | C6H3Cl2NO | 176.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA395790250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS98-08-8
IUPAC Name(trifluoromethyl)benzene
Molecular FormulaC7H5F3
InChI KeyGETTZEONDQJALK-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
Refractive index1.4130 to 1.4150 (20°C, 589 nm)