Di(ethylene glycol) divinyl ether, 95%
Di(ethylene glycol) divinyl ether, 95%
Di(ethylene glycol) divinyl ether, 95%
Di(ethylene glycol) divinyl ether, 95%
Thermo Scientific Chemicals
Di(ethylene glycol) divinyl ether, 95%
CAS: 764-99-8 | C8H14O3 | 158.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA39541050050 mLGlass bottle
Catalog number FSA395410500
Price (MYR)
1,397.00
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,397.00
Add to cart
Specifications
Chemical Name or MaterialDi(ethylene glycol) divinyl ether
CAS764-99-8
Melting Point-21.0°C
Density0.9700g/mL
EINECS Number212-133-3