Tetrabutylphosphonium hydroxide, 40 wt.% solution in water
Tetrabutylphosphonium hydroxide, 40 wt.% solution in water
Tetrabutylphosphonium hydroxide, 40 wt.% solution in water
Tetrabutylphosphonium hydroxide, 40 wt.% solution in water
Thermo Scientific Chemicals
Tetrabutylphosphonium hydroxide, 40 wt.% solution in water
CAS: 14518-69-5 | C16H37OP | 276.45 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA395031000100 mLPlastic Bottle
Catalog number FSA395031000
Price (MYR)
1,075.00
Add to cart
Quantity:
100 mL
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,075.00
Add to cart
Chemical Identifiers
CAS100-02-7
IUPAC Name4-nitrophenol
Molecular FormulaC6H5NO3
InChI KeyBTJIUGUIPKRLHP-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to brown
Assay (GC)≥98.5%
Melting Point (clear melt)108-116?C
FormCrystals, crystalline/granular powder and/or chunks
Identification (FTIR)Conforms
For C-C Bond Formation (Olefination)
RUO – Research Use Only