Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Plastic bottle
Catalog number FSA393320010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS3115-68-2
IUPAC Nametetrabutylphosphanium bromide
Molecular FormulaC16H36BrP
InChI KeyRKHXQBLJXBGEKF-UHFFFAOYSA-M
SMILES[Br-].CCCC[P+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)100.0-106.0?C
Water Content (Karl Fischer Titration)≤0.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Titration ex Bromide)≥98.5 to ≤101.5%
View more
Suitable for battery materials development.
For Manufacturing and Laboratory Use. Not for beverage use