2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
CAS: 7305-71-7 | C4H6N2S | 114.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA392790250
Price (MYR)
467.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
467.00
Add to cart
Chemical Identifiers
CAS90-64-2
IUPAC Name2-hydroxy-2-phenylacetic acid
Molecular FormulaC8H8O3
InChI KeyIWYDHOAUDWTVEP-UHFFFAOYNA-N
SMILESOC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Titration with NaOH>=99.0 %
Appearance (Form)Crystalline powder or crystals
Melting point117°C to 121°C