2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
CAS: 7305-71-7 | C4H6N2S | 114.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA392790050
Price (MYR)
235.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
235.00
Add to cart
Chemical Identifiers
CAS372-31-6
IUPAC Nameethyl 4,4,4-trifluoro-3-oxobutanoate
Molecular FormulaC6H7F3O3
InChI KeyOCJKUQIPRNZDTK-UHFFFAOYSA-N
SMILESCCOC(=O)CC(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.3750 to 1.3780 (20°C, 589 nm)