2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized, ACROS Organics, Quantity: 10L, Packaging: Metal Drum, Boiling Point: 78.0 deg.C to 80.0 deg.C, CAS: 96-47-9, Colorless, Grade: For Grignard reaction, Melting Point: -136.0 deg.C, Molecular Formula: C5H10O, Molecular Weight: 86.13
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number FSA392380025
Price (MYR)
1,841.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,841.00
Add to cart
Chemical Identifiers
CAS95-03-4
IUPAC Name5-chloro-2-methoxyaniline
Molecular FormulaC7H8ClNO
InChI KeyWBSMIPLNPSCJFS-UHFFFAOYSA-N
SMILESCOC1=CC=C(Cl)C=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown to dark brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)77.5-83.5?C