Yttrium acetate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals
Yttrium acetate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals
Yttrium acetate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals
Yttrium acetate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals
Alfa Aesar
Yttrium acetate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals
CAS: 207801-28-3 | C6H9O6Y | 266.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA392210250
Price (MYR)
134.01
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
134.01
Add to cart
Chemical Identifiers
CAS19004-19-4
IUPAC Namedicopper(2+) pentahydrate tetranitrate
Molecular FormulaCu2H10N4O17
InChI KeyIZDJJEMZQZQQQQ-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue
FormCrystalline powder or lumpy powder
Assay (unspecified)>97.5%
Residual water17.8-22.4% Water
for ultra trace analysis
RUO – Research Use Only