N,N-Dimethylacetamide, 99.5+%, for analysis
N,N-Dimethylacetamide, 99.5+%, for analysis
N,N-Dimethylacetamide, 99.5+%, for analysis
N,N-Dimethylacetamide, 99.5+%, for analysis
Alfa Aesar
N,N-Dimethylacetamide, 99.5+%, for analysis
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Catalog number FSA390800010
Price (MYR)
546.00
Add to cart
Request bulk or custom format
Price (MYR)
546.00
Add to cart
Chemical Identifiers
CAS54827-17-7
IUPAC Name3,3',5,5'-tetramethyl-[1,1'-biphenyl]-4,4'-diamine
Molecular FormulaC16H20N2
InChI KeyUAIUNKRWKOVEES-UHFFFAOYSA-N
SMILESCC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HClO498.0 to 102.0 %
Infrared spectrumConforms
HPLC>=99.0 %
Sulfated ash=<0.2 %
View more
For analysis
RUO – Research Use Only