Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Chloroform, >-99%, CHCl3, CAS Number-67-66-3, 64-17-5, trichloromethane, trichloroform, trichlormethan, methyl trichloride, cloroformio, methenyl trichloride, formyl trichloride, methane, trichloro, chloroforme, methane trichloride, 2.5L
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA390760010
Price (MYR)
587.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
587.00
Add to cart
Specifications
Chemical Name or MaterialChloroform
Melting Point-63.0°C
Name NoteStabilized with ethanol
Absorbance0.009 max. at 300nm, 0.046 max. at 265nm, 0.301 max. at 250nm, 0.699 max. at 245nm, 1 cm cell vs water, 1 cm cell vs water at 0nm
CAS67-66-3
View more
For HPLC
RUO – Research Use Only