tert-Butanol, Thermo Scientific Chemicals
tert-Butanol, Thermo Scientific Chemicals
tert-Butanol, Thermo Scientific Chemicals
tert-Butanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butanol, Thermo Scientific Chemicals
CAS: 75-65-0 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic Bottle
Catalog number FSA390690025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS540-69-2
IUPAC Nameformic acid amine
Molecular FormulaCH5NO2
InChI KeyVZTDIZULWFCMLS-UHFFFAOYSA-N
SMILESN.OC=O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMaterial sourced in UK and US
Identification (FTIR)Conforms (UK-sourced material)
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0% (UK-sourced material)