1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
CAS: 872-50-4 | C5H9NO | 99.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Packaging:
Glass Bottle
Catalog number FSA390680025
Price (MYR)
1,250.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,250.00
Add to cart
Specifications
Chemical Name or Material1-Methyl-2-pyrrolidinone
Melting Point-24.0°C
CAS872-50-4
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May damage the unborn child.
View more
Suitable for battery materials development.
RUO – Research Use Only