Iron(III) phosphate hydrate, extra pure
Iron(III) phosphate hydrate, extra pure
Iron(III) phosphate hydrate, extra pure
Iron(III) phosphate hydrate, extra pure
Thermo Scientific Chemicals
Iron(III) phosphate hydrate, extra pure
CAS: 51833-68-2 | FeH2O5P | 168.83 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA389955000500 gGlass bottle
Catalog number FSA389955000
Price (MYR)
765.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
765.00
Add to cart
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
Diameter0.45-0.55mm
Total Metal Impurities≤ 0.001%
CommentGold
FormWire