Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water, Thermo Scientific Chemicals
Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water, Thermo Scientific Chemicals
Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water, Thermo Scientific Chemicals
Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water, Thermo Scientific Chemicals
CAS: 124-64-1 | C4H12ClO4P | 190.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA389448000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
800 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS121-44-8
IUPAC Nametriethylamine
Molecular FormulaC6H15N
InChI KeyZMANZCXQSJIPKH-UHFFFAOYSA-N
SMILESCCN(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<15 APHA
Infrared spectrumConforms
GC>=98.5 %
Water=<0.2 % (K.F.)
View more
for C-C Bond Formation (Olefination)
RUO – Research Use Only