Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water, Thermo Scientific Chemicals
Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water, Thermo Scientific Chemicals
Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water, Thermo Scientific Chemicals
Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water, Thermo Scientific Chemicals
CAS: 124-64-1 | C4H12ClO4P | 190.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
Glass bottle
Catalog number FSA389441000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS119-64-2
IUPAC Name1,2,3,4-tetrahydronaphthalene
Molecular FormulaC10H12
InChI KeyCXWXQJXEFPUFDZ-UHFFFAOYSA-N
SMILESC1CCC2=CC=CC=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Color (APHA)≤20 (US sourced material)
Assay (GC)≥96.0% (Non-US sourced material)
Assay from Supplier's CofA≤96.0% (US sourced material)
View more
for C-C Bond Formation (Olefination)
RUO – Research Use Only