n-Pentane, 99+%, Thermo Scientific Chemicals
n-Pentane, 99+%, Thermo Scientific Chemicals
n-Pentane, 99+%, Thermo Scientific Chemicals
n-Pentane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Pentane, 99+%, Thermo Scientific Chemicals
CAS: 109-66-0 | C5H12 | 72.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA389070010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Pentane
Melting Point-130.0°C
Absorbance(1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.03 max. at 230nm, 0.2 max. at 210nm, 0.75 max. at 200nm
CAS109-66-0
ColorColorless
View more
For HPLC
RUO – Research Use Only