Phosphoric acid, pure, Thermo Scientific Chemicals
Phosphoric acid, pure, Thermo Scientific Chemicals
Phosphoric acid, pure, Thermo Scientific Chemicals
Phosphoric acid, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphoric acid, pure, Thermo Scientific Chemicals
CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA389021000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4023-65-8
IUPAC Name(1E)-prop-1-ene-1,2,3-tricarboxylic acid
Molecular FormulaC6H6O6
InChI KeyGTZCVFVGUGFEME-HNQUOIGGSA-N
SMILESOC(=O)CC(=C/C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5% (non-U.S. specification)
Proton NMRConforms (U.S. specification)
Assay (HPLC)≥97.5% (non-U.S. specification)