D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP, Thermo Scientific Chemicals
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP, Thermo Scientific Chemicals
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP, Thermo Scientific Chemicals
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP, Thermo Scientific Chemicals
Thermo Scientific Chemicals
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP, Thermo Scientific Chemicals
CAS: 50-99-7 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number FSA388190010
Price (MYR)
399.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
399.00
Add to cart
Chemical Identifiers
CAS56375-79-2
IUPAC Nametributyl(methyl)azanium iodide
Molecular FormulaC13H30IN
InChI KeyDXJLCRNXYNRGRA-UHFFFAOYSA-M
SMILES[I-].CCCC[N+](C)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4550-1.4590 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (Titration ex Chloride)>73.0 to <78.0%