Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals
Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals
Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals
Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals
CAS: 82113-65-3 | C2HF6NO4S2 | 281.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number FSA387600050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1572-93-6
IUPAC Name3-methylbutan-2-ol
Molecular FormulaC5H12O
InChI KeyMXLMTQWGSQIYOW-UHFFFAOYNA-N
SMILESCC(C)C(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Optical Rotation-4.8 ± 0.1? (neat)
Refractive Index1.4075-1.4120 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Coupling reagent
RUO – Research Use Only