Boiling Ston
Thermo Scientific Chemicals
Boiling Ston
For analytical work
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
200 g
Catalog number FSA386200500
Price (MYR)
231.00
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
231.00
Add to cart
Chemical Identifiers
CAS110-01-0
IUPAC Namethiolane
Molecular FormulaC4H8S
InChI KeyRAOIDOHSFRTOEL-UHFFFAOYSA-N
SMILESC1CCSC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5030-1.5070 @ 20°C