Glass Wool
Thermo Scientific Chemicals
Glass Wool
Filter aid
Have Questions?
Change viewbuttonViewtableView
Quantity:
50g
250g
1kg
Catalog number FSA386060500
Price (MYR)
247.00
Add to cart
Quantity:
50g
Request bulk or custom format
Price (MYR)
247.00
Add to cart
Chemical Identifiers
CAS153624-46-5
IUPAC Name[4-(propan-2-yloxy)phenyl]boronic acid
Molecular FormulaC9H13BO3
InChI KeyCJUHQADBFQRIMC-UHFFFAOYSA-N
SMILESCC(C)OC1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%