Calcium hydroxide, 99.995%, (trace metal basis)
Calcium hydroxide, 99.995%, (trace metal basis)
Calcium hydroxide, 99.995%, (trace metal basis)
Calcium hydroxide, 99.995%, (trace metal basis)
Thermo Scientific Chemicals
Calcium hydroxide, 99.995%, (trace metal basis)
CAS: 1305-62-0 | CaH2O2 | 74.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3856100505 gGlass bottle
Catalog number FSA385610050
Price (MYR)
638.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
638.00
Add to cart
Chemical Identifiers
CAS2142-63-4
IUPAC Name1-(3-bromophenyl)ethan-1-one
Molecular FormulaC8H7BrO
InChI KeyJYAQYXOVOHJRCS-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5749 to 1.5769 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
for ultra trace analysis
RUO – Research Use Only