(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
CAS: 18107-18-1 | C4H10N2Si | 114.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA385330250
Price (MYR)
1,527.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,527.00
Add to cart
Chemical Identifiers
CAS75-52-5
IUPAC Namenitromethane
Molecular FormulaCH3NO2
InChI KeyLYGJENNIWJXYER-UHFFFAOYSA-N
SMILESC[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.0%
Refractive Index1.3800-1.3840 @ 20?C
Alkylation reagent
RUO – Research Use Only