(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
CAS: 18107-18-1 | C4H10N2Si | 114.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA385330050
Price (MYR)
504.00
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
504.00
Add to cart
Chemical Identifiers
CAS1436-34-6
IUPAC Name2-butyloxirane
Molecular FormulaC6H12O
InChI KeyWHNBDXQTMPYBAT-UHFFFAOYNA-N
SMILESCCCCC1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4040 to 1.4070 (20°C, 589 nm)
Alkylation reagent
RUO – Research Use Only