Osmium 99.9%, Thermo Scientific Chemicals
Osmium 99.9%, Thermo Scientific Chemicals
Osmium 99.9%, Thermo Scientific Chemicals
Osmium 99.9%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Osmium 99.9%, Thermo Scientific Chemicals
CAS: 7440-04-2 | Os | 190.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA385160010
Price (MYR)
1,365.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,365.00
Add to cart
Chemical Identifiers
CAS328-80-3
IUPAC Name3-nitro-5-(trifluoromethyl)benzoate
Molecular FormulaC8H3F3NO4
InChI KeyODCLHXGXGFBBTA-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC(=CC(=C1)C(F)(F)F)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (Silylated GC)≥96.0%
Appearance (Color)Pale cream to pale yellow
Melting Point (clear melt)125.0-135.0?C
for ultra trace analysis
RUO – Research Use Only