n-Hexane, for analysis, contains appr. 95% n-Hexane, Thermo Scientific Chemicals
n-Hexane, for analysis, contains appr. 95% n-Hexane, Thermo Scientific Chemicals
n-Hexane, for analysis, contains appr. 95% n-Hexane, Thermo Scientific Chemicals
n-Hexane, for analysis, contains appr. 95% n-Hexane, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Hexane, for analysis, contains appr. 95% n-Hexane, Thermo Scientific Chemicals
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
25 L
Catalog number FSA383800025
Price (MYR)
536.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
536.00
Add to cart
Specifications
Chemical Name or Materialn-Hexane
Melting Point-95.0°C
CAS110-54-3
Health Hazard 1Danger
Health Hazard 2GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor.
View more
For analysis
RUO – Research Use Only