Dichloromethane 99+%, stabilized with amylene, pure, Thermo Scientific Chemicals
Dichloromethane 99+%, stabilized with amylene, pure, Thermo Scientific Chemicals
Dichloromethane 99+%, stabilized with amylene, pure, Thermo Scientific Chemicals
Dichloromethane 99+%, stabilized with amylene, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloromethane 99+%, stabilized with amylene, pure, Thermo Scientific Chemicals
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
Catalog number FSA383780250
Price (MYR)
1,873.00
Add to cart
Quantity:
25 L
Request bulk or custom format
Price (MYR)
1,873.00
Add to cart
Chemical Identifiers
CAS7761-88-8
IUPAC Namesilver(1+) nitrate
Molecular FormulaAgNO3
InChI KeySQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES[Ag+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Copper (Cu)=<1 ppm
Loss on drying=<0.1 % (1 g, 70°C, 2 h)
Chloride (Cl)=<5 ppm