4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals
4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals
4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals
4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals
CAS: 3731-53-1 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number FSA382080250
Price (MYR)
347.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
347.00
Add to cart
Chemical Identifiers
CAS2052-49-5
IUPAC Nametetrabutylazanium hydroxide
Molecular FormulaC16H37NO
InChI KeyVDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES[OH-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HCl0.95 to 1.10 M
Appearance (Color)Clear colorless to yellow
Appearance (Form)Solution