Piperine, 98%, Thermo Scientific Chemicals
Piperine, 98%, Thermo Scientific Chemicals
Piperine, 98%, Thermo Scientific Chemicals
Piperine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Piperine, 98%, Thermo Scientific Chemicals
CAS: 94-62-2 | C17H19NO3 | 285.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA381450050
Price (MYR)
397.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
397.00
Add to cart
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 1.0mm (0.04in) diameter.
Assay from Supplier's CofA≥99.9996% metals basis