Lithium Bis(trifluoromethanesulfonimide), 99%, Thermo Scientific Chemicals
Lithium Bis(trifluoromethanesulfonimide), 99%, Thermo Scientific Chemicals
Lithium Bis(trifluoromethanesulfonimide), 99%, Thermo Scientific Chemicals
Lithium Bis(trifluoromethanesulfonimide), 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lithium Bis(trifluoromethanesulfonimide), 99%, Thermo Scientific Chemicals
CAS: 90076-65-6 | C2F6LiNO4S2 | 287.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA381030100
Price (MYR)
792.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
792.00
Add to cart
Chemical Identifiers
CAS5434-21-9
IUPAC Name4-aminobenzene-1,2-dicarboxylic acid
Molecular FormulaC8H7NO4
InChI KeyOXSANYRLJHSQEP-UHFFFAOYSA-N
SMILESNC1=CC=C(C(O)=O)C(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to orange to cream to pale brown
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%