4-Ethynylanisole, 97%, Thermo Scientific Chemicals
4-Ethynylanisole, 97%, Thermo Scientific Chemicals
4-Ethynylanisole, 97%, Thermo Scientific Chemicals
4-Ethynylanisole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Ethynylanisole, 97%, Thermo Scientific Chemicals
CAS: 768-60-5 | C9H8O | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA379920010
Price (MYR)
420.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
420.00
Add to cart
Chemical Identifiers
CAS89-83-8
IUPAC Name5-methyl-2-(propan-2-yl)phenol
Molecular FormulaC10H14O
InChI KeyMGSRCZKZVOBKFT-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(C)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point48°C to 52°C
GC>=98.5 %