tert-Butyl methyl ether, 99.9%, extra pure, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99.9%, extra pure, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99.9%, extra pure, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99.9%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butyl methyl ether, 99.9%, extra pure, Thermo Scientific Chemicals
CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA378720010
Price (MYR)
355.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
355.00
Add to cart
Chemical Identifiers
CAS15708-41-5
IUPAC Nameiron(3+) sodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Molecular FormulaC10H12FeN2NaO8
InChI KeyMKWYFZFMAMBPQK-UHFFFAOYSA-J
SMILES[Na+].[Fe+3].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)Total EDTA: ≥65.5%
Formpowder
Assay (Titration)Fe (III) 12.5 - 13.5%
Appearance (Color)Pale yellow