N,N-Dimethylacetamide, 99.5%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.5%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.5%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.5%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.5%, extra pure, Thermo Scientific Chemicals
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA378710010
Price (MYR)
432.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
432.00
Add to cart
Chemical Identifiers
CAS7446-70-0
IUPAC Namealuminium(3+) trichloride
Molecular FormulaAlCl3
InChI KeyVSCWAEJMTAWNJL-UHFFFAOYSA-K
SMILES[Al+3].[Cl-].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Metal Impurities0.01% max.
Appearance (Color)White
Residual water< 100 ppm