Calcium phosphate, monobasic, 90%, pure
Calcium phosphate, monobasic, 90%, pure
Calcium phosphate, monobasic, 90%, pure
Calcium phosphate, monobasic, 90%, pure
Thermo Scientific Chemicals
Calcium phosphate, monobasic, 90%, pure
CAS: 7758-23-8 | CaH4O8P2 | 234.05 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3786100101 kgPlastic bottle
Catalog number FSA378610010
Price (MYR)
588.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
588.00
Add to cart
Specifications
Chemical Name or MaterialCalcium phosphate, monobasic
CAS7758-23-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye damage.
Health Hazard 3GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici