Methyl-beta-cyclodextrin, average Mw 1310, Thermo Scientific Chemicals
Methyl-beta-cyclodextrin, average Mw 1310, Thermo Scientific Chemicals
Methyl-beta-cyclodextrin, average Mw 1310, Thermo Scientific Chemicals
Methyl-beta-cyclodextrin, average Mw 1310, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl-beta-cyclodextrin, average Mw 1310, Thermo Scientific Chemicals
CAS: 128446-36-6 | C54H94O35 | 1303.311 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA377110050
Price (MYR)
293.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
293.00
EA
Add to cart
Chemical Identifiers
CAS19756-04-8
Molecular FormulaC8H24N4Zr
InChI KeyDWCMDRNGBIZOQL-UHFFFAOYSA-N
SMILES[Zr+4].C[N-]C.C[N-]C.C[N-]C.C[N-]C
Molecular Weight (g/mol)267.53
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline solid
NMR (1H-NMR)Consistent with structure
Appearance (Color)Yellow-green
Chelation/Complexation reagent
RUO – Research Use Only