DL-Lactide, 99%, Thermo Scientific Chemicals
DL-Lactide, 99%, Thermo Scientific Chemicals
DL-Lactide, 99%, Thermo Scientific Chemicals
DL-Lactide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Lactide, 99%, Thermo Scientific Chemicals
CAS: 95-96-5 | C6H8O4 | 144.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA376850250
Price (MYR)
604.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
604.00
Add to cart
Chemical Identifiers
CAS1638-86-4
IUPAC Namediethyl phenylphosphonite
Molecular FormulaC10H15O2P
InChI KeyRVDJLKVICMLVJQ-UHFFFAOYSA-N
SMILESCCOP(OCC)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.5085-1.5125 @ 20?C