p-Toluidine, 99%, crystalline molten mass, Thermo Scientific Chemicals
p-Toluidine, 99%, crystalline molten mass, Thermo Scientific Chemicals
p-Toluidine, 99%, crystalline molten mass, Thermo Scientific Chemicals
p-Toluidine, 99%, crystalline molten mass, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluidine, 99%, crystalline molten mass, Thermo Scientific Chemicals
CAS: 106-49-0 | C7H9N | 107.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
View all
Glass bottle
Catalog number FSA376350010
Price (MYR)
540.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
540.00
Add to cart
Specifications
Chemical Name or Materialp-Toluidine
Name Note99%
CAS106-49-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic in contact with skin.
Suspected of causing cancer.
Causes serious eye irritation.
May cause an allergic skin reaction.
Toxic if inhaled.
Very toxic to aquatic life.