4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals
4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals
4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals
4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals
CAS: 1074-36-8 | C7H6O2S | 154.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA373300050
Price (MYR)
614.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
614.00
Add to cart
Chemical Identifiers
CAS5990-32-9
IUPAC Nametrizinc(2+) bis(2-hydroxypropane-1,2,3-tricarboxylate) dihydrate
Molecular FormulaC12H14O16Zn3
InChI KeyOXAGUGIXGVHDGD-UHFFFAOYSA-H
SMILESO.O.[Zn++].[Zn++].[Zn++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Zn 31% min