Phenyltrimethoxysilane, 85%, Thermo Scientific Chemicals
Phenyltrimethoxysilane, 85%, Thermo Scientific Chemicals
Phenyltrimethoxysilane, 85%, Thermo Scientific Chemicals
Phenyltrimethoxysilane, 85%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyltrimethoxysilane, 85%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass bottle
Catalog number FSA370642500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.