Tributylphenyltin, Thermo Scientific Chemicals
Tributylphenyltin, Thermo Scientific Chemicals
Tributylphenyltin, Thermo Scientific Chemicals
Tributylphenyltin, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tributylphenyltin, Thermo Scientific Chemicals
CAS: 960-16-7 | C18H32Sn | 367.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
50 g
Packaging:
Glass bottle
Catalog number FSA370230010
Price (MYR)
281.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
281.00
Add to cart
Chemical Identifiers
CAS628-77-3
IUPAC Name1,5-diiodopentane
Molecular FormulaC5H10I2
InChI KeyIAEOYUUPFYJXHN-UHFFFAOYSA-N
SMILESICCCCCI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless or yellow to orange
Refractive Index1.5980 - 1.6040 @20?C
Formliquid
Assay (GC)> 96.0%