3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
CAS: 107-86-8 | C5H8O | 84.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA370180250
Price (MYR)
1,175.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,175.00
Add to cart
Chemical Identifiers
CAS7389-87-9
IUPAC Namemethyl 2-amino-3-(1H-imidazol-5-yl)propanoate hydrochloridyl
Molecular FormulaC7H11ClN3O2
InChI KeyVXXIPPPPKNWFLK-UHFFFAOYNA-N
SMILES[Cl].COC(=O)C(N)CC1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Appearance (Color)White
Optical Rotation10.0 ± 1.0? (C=2 in water)