3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
CAS: 107-86-8 | C5H8O | 84.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA370180050
Price (MYR)
499.00
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
499.00
Add to cart
Chemical Identifiers
CAS88-75-5
IUPAC Name2-nitrophenol
Molecular FormulaC6H5NO3
InChI KeyIQUPABOKLQSFBK-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or lumps or fused solid or flakes
Identification (FTIR)Conforms
Appearance (Color)Yellow to green to brown
Melting Point (clear melt)42.0-49.0?C