2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals
2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals
2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals
2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals
CAS: 31106-82-8 | C6H6BrN·HBr | 252.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA370110050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-(Bromomethyl)pyridine hydrobromide
CAS31106-82-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0