Aminoguanidine hydrochloride, 98%, Thermo Scientific Chemicals
Aminoguanidine hydrochloride, 98%, Thermo Scientific Chemicals
Aminoguanidine hydrochloride, 98%, Thermo Scientific Chemicals
Aminoguanidine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aminoguanidine hydrochloride, 98%, Thermo Scientific Chemicals
CAS: 1937-19-5 | CH6N4·HCl | 110.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA368910250
Price (MYR)
370.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
370.00
Add to cart
Chemical Identifiers
CAS1633-22-3
IUPAC Nametricyclo[8.2.2.2⁴,⁷]hexadeca-1(12),4,6,10,13,15-hexaene
Molecular FormulaC16H16
InChI KeyOOLUVSIJOMLOCB-UHFFFAOYSA-N
SMILESC1CC2=CC=C(CCC3=CC=C1C=C3)C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.5%
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale yellow