Glycerine trioleate, 98%, Thermo Scientific Chemicals
Glycerine trioleate, 98%, Thermo Scientific Chemicals
Glycerine trioleate, 98%, Thermo Scientific Chemicals
Glycerine trioleate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycerine trioleate, 98%, Thermo Scientific Chemicals
CAS: 122-32-7 | C57H104O6 | 885.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 mL
5 mL
25 mL
Catalog number FSA368121000
Price (MYR)
15,054.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
15,054.00
Add to cart
Chemical Identifiers
CAS2893-78-9
IUPAC Namesodium 3,5-dichloro-2,4,6-trioxo-1,3,5-triazinan-1-ide
Molecular FormulaC3Cl2N3NaO3
InChI KeyMSFGZHUJTJBYFA-UHFFFAOYSA-M
SMILES[Na+].ClN1C(=O)[N-]C(=O)N(Cl)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystals or granules
Infrared spectrumConforms
Titration with HClO4>=97.5 % (On dry substance)
Loss on drying=<5 %