Glycerine trioleate, 98%, Thermo Scientific Chemicals
Glycerine trioleate, 98%, Thermo Scientific Chemicals
Glycerine trioleate, 98%, Thermo Scientific Chemicals
Glycerine trioleate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycerine trioleate, 98%, Thermo Scientific Chemicals
CAS: 122-32-7 | C57H104O6 | 885.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mL
5 mL
25 mL
100 mL
Catalog number FSA368120010
Price (MYR)
481.00
Add to cart
Quantity:
1 mL
Request bulk or custom format
Price (MYR)
481.00
Add to cart
Chemical Identifiers
CAS10417-94-4
IUPAC Name(5Z,8Z,11Z,14Z,17Z)-icosa-5,8,11,14,17-pentaenoic acid
Molecular FormulaC20H30O2
InChI KeyJAZBEHYOTPTENJ-JLNKQSITSA-N
SMILESCC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Assay (unspecified)>98%
FormOil